Disposable earplugs, uvex x-fit
Catalog # 111-2345
Supplier: UVEX
Specifications
- Colour:Bright Green
- Description:Earplugs, without cord, bagged in pairs
- Seq:01
- Environmentally Preferable:
- Pk:200 PAIR
Specifications
About this item
The ergonomically shaped x-fit disposable earplug is characterised by very high sound absorption levels, making it suitable for use in very loud environments. The x-fit foam earplug has a sealed surface, preventing foreign bodies from penetrating the plug.
- Soft foam offers real comfort, even when worn for extended periods
- Ergonomic shape adapts to the natural shape of the ear canal
- 'x' emboss in the end of the plug makes it easy to remove while simultaneously reducing the pressure on the outer wall of the ear canal
- Easy insertion and removal
- For medium to large ear canals
- High visibility colour for easy visual recognition
SNR: 37 dB
Certifications: EN 352-2:2002