ContactUsLinkComponent
UV Meters, analog, J-221 & J-225
UV Meters, analog, J-221 & J-225
Certificates
undefined
UV Meters, analog, J-221 & J-225
Specifications
  • Catalog No:
  • UVPA97-0003-01
  • UVPA97-0004-01
  • Model:
  • J221
  • J225
  • Measuring range:
  • 300 - 400 nm
  • 220 - 280 nm
  • Sensor:
  • built-in-sensor
  • built-in-sensor
  • Display:
  • Attenuation screen, 5X
  • Attenuation screen, 5X
  • W×D×H:
  • 76×76×76 mm
  • 76×76×76 mm
  • Weight:
  • 250 g
  • 250 g