Substrate for the detection of immobilised HRP
Katalognr. 732-1208
Anbieter: Uptima
CAS-Nummer:
Technische Daten
- Pk:1 KIT
- Description:UptiLight
Technische Daten
Über diesen Artikel
Substrate optimised for the detection of immobilised HRP in Western, Southern, Northern and dot blot applications.
- Compatibility with nylon, nitrocellulose and PVDF membranes
- Fast response time - specific detection in 1 to 10 minutes
- High sensitivity - detects up to 1 to 2 pg antigen
- Long signal duration - stable reagent allows repeated exposure