Dip slides
About this item
Two-sided slide with agar-agar centre. Operating instructions: 1. Unscrew the cap containing the agar-agar slide. Hold the bottle in the other hand and lightly apply pressure to the sampling area using the agar-agar slide; or 2. Apply directly to the middle of the swab brush if you have completed a swab sample; or 3. Immerse the agar-agar slide in the liquid to analyse. 4. Insert the slide into the bottle and screw back on. Place in the oven at 37 °C for 24/48 hours. To detect yeasts, increase the incubation by 24/48 hours at 25/30 °C.
- Box of 10 slides with separate labels.
- Use-by date: six months after manufacture
- Storage: ambient temperature (max. 25 °C)
5 Options Available Below
Product Details & Documents
Two-sided slide with agar-agar centre. Operating instructions: 1. Unscrew the cap containing the agar-agar slide. Hold the bottle in the other hand and lightly apply pressure to the sampling area using the agar-agar slide; or 2. Apply directly to the middle of the swab brush if you have completed a swab sample; or 3. Immerse the agar-agar slide in the liquid to analyse. 4. Insert the slide into the bottle and screw back on. Place in the oven at 37 °C for 24/48 hours. To detect yeasts, increase the incubation by 24/48 hours at 25/30 °C.
- Box of 10 slides with separate labels.
- Use-by date: six months after manufacture
- Storage: ambient temperature (max. 25 °C)
Documents
Product Family Options
Product Information
- PkAvailabilityPrice
- LIOF525622Flexible agar-agar contact slides for total flora/coliformsFlexible agar-agar contact slides for total flora/coliforms20Log in for availability1 * 20 STSFr. 75.70
- LIOF525282Flexible agar-agar contact slides for pseudomonas
- LIOF525302Flexible agar-agar contact slides for total flora
- LIOF525312Flexible agar-agar contact slides for enterobacter
- LIOF525272Flexible agar-agar contact slides for total flora/yeasts and moulds plus neutraliserFlexible agar-agar contact slides for total flora/yeasts and moulds plus neutraliser20Log in for availability1 * 20 STSFr. 55.10