Cell Recovery Solution, Corning®
Corning® Cell Recovery Solution, 100 mL
Supplier: Corning
Cell Recovery Solution, Corning®
Catalog # 734-0107
Supplier: Corning
CAS Number:
Jump to:
Product Details & Documents
Corning® Cell Recovery Solution allows for the recovery of cells cultured on Corning® Matrigel® Basement Membrane Matrix for subsequent biochemical analysis. Corning® Cell Recovery Solution depolymerises Corning® Matrigel® Matrix gels without enzymatic digests and lengthy incubation periods at high temperatures. Cells are released without damage thereby avoiding biochemical changes during incubation and digestion of extracellular portions of cell-surface receptors and adhesion molecules.
- Non-enzymatic proprietary solution
- Tested for ability to depolymerise a 1 mm thick layer of gelled BD Matrigel™ matrix after one hour at 2 to 8 °C
- Filtered and tested and found negative for bacteria, fungi, and mycoplasma
- Stable for at least three months at 2 to 8 °C
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:100 mL
- Description:Corning® Cell Recovery Solution
- Cat. No.:734-0107
- Environmental certification:ISO 14001 - Bedford
- Environmentally Preferable:Low Manufacturing Impact



