ContactUsLinkComponent
Pipette stand for single channel pipettes, WHEATON®
Pipette stand for single channel pipettes, WHEATON®
Catalog # WHEAW851361
Supplier:  DWK Life Sciences
undefined
Pipette stand for single channel pipettes, WHEATON®
Catalog # WHEAW851361
Supplier:  DWK Life Sciences
Specifications
  • For:
    7 single channel pipettes
  • Colour:
    Light grey
  • Description:
    Pipettor universal workstation, model 337, with anti-slip pads
  • Pk:
    1
Specifications
About this item
This space-saving pipette stand holds up to seven single channel pipettes of most brands.
  • Made of sturdy polyamid material, easy to clean
  • Anti-slip pads guarantee bench stability
  • Work station fits new and old generations of Socorex and other brand pipettes