VWR®, folding linen tester
Katalognr. 630-1087
Anbieter: VWR Collection
Technische Daten
- Ø:10 mm
- Magnification:10×
- Description:Folding magnifier, plastic
- Seq:02
- Pk:1
Technische Daten
Über diesen Artikel
These linen testers are widely used in industrial electronics and laboratories, as well as for photography.
- Glass lens
- Different sizes and magnifications
- Folding instruments
Colour: Black