VWR®, Accessories for Block Heaters Analogue/Digital
Plexiglas® covers with 2 open sides reduce airflow, providing additional temperature stability in low temperature applications.
Frequently Bought Together
Plexiglas® covers with 2 open sides reduce airflow, providing additional temperature stability in low temperature applications.