ContactUsLinkComponent
Viewing cabinets, Chromato-Vue®, C-75G
Viewing cabinets, Chromato-Vue®, C-75G
  UVPA95-0253-02
undefined
Viewing cabinets, Chromato-Vue®, C-75G
  UVPA95-0253-02
  • Model:
    C-75
  • Wavelength:
    254/365 nm
  • Description:
    Chromato-Vue® C-75, high performance, with two UV tubes
  • Output:
    2×15 W
  • Pk:
    1
  • For:
    Fluorescence studies and non destructive testing