Vials with insert, WHEATON®
Fournisseur: DWK Life Sciences
Certificats
À propos de cet article
These vials made of PP uses either aluminum crimp seals or plastic snap cap closures, which eliminate the need for crimping and decapping tools, and provide a larger target area.
- Available with snap/crimp or screw top finish
- Unique cone-shaped bottom ensures maximum sample retrieval
Conditionnement: Packaged as 100 vials per shelf pack or 1000 vials per case.
Frequently Bought Together