Orders LinkContactUsLinkComponent
Washers, BOLA
Washers, BOLA
Supplier:   Bohlender
Washers, BOLA
Supplier:  Bohlender
Certificates
Washers, BOLA
4 Options Available Below
Product Details & Documents
PTFE, white.
  • Temperature resistance: from −200 to +250 °C
  • Universal chemical resistance
  • FDA conform
  • Similar to DIN 125-1
Product Family Options
Product Information
  • Ø
    Availability
    Price
  • BOHLH1126-26
    M 8 thread
    M 8 thread
    16 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 3.80
    SFr. 3.80
    1 * 1 ST
       SFr. 3.80
  • BOHLH1126-22
    M 6 thread
    M 6 thread
    12 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 4.80
    SFr. 4.80
    1 * 1 ST
       SFr. 4.80
  • BOHLH1126-18
    M 5 thread
    M 5 thread
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 4.00
    SFr. 4.00
    1 * 1 ST
       SFr. 4.00
  • BOHLH1126-14
    M 4 thread
    M 4 thread
    9 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 3.90
    SFr. 3.90
    1 * 1 ST
       SFr. 3.90
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.