Staining dishes, WHEATON®
Soda-lime glass and stainless steel.
- Handle permanently attached
- Hinged closure
- Easy insertion and removal of slides
The slotted rack hold 50 microscope slides 75×25 mm, 75×38 mm or 75×51 mm. Rack is made of non tarnishable, stainless steel that is resistant to staining solutions.
Soda-lime glass and stainless steel.
- Handle permanently attached
- Hinged closure
- Easy insertion and removal of slides
The slotted rack hold 50 microscope slides 75×25 mm, 75×38 mm or 75×51 mm. Rack is made of non tarnishable, stainless steel that is resistant to staining solutions.
Product Family Options
- DescriptionW×D×H
- Staining dish complete, includes dish, cover and rack185×88×68 mm1 * 6 STSFr. 2’260.00
- Staining racks with handle for 50 slides- mm1 * 3 STSFr. 363.00