Order Entry
Switzerland
ContactUsLinkComponent
 
Petri dishes, square with grid
Catalog # 391-3555
Supplier:  PHOENIX BIOMEDICAL
undefined
Petri dishes, square with grid
Catalog # 391-3555
Supplier:  PHOENIX BIOMEDICAL

Specifications

  • Ø:
    100 mm
  • Packed:
    300 per case (15 sleeves)
  • Sterility:
    Non sterile
  • Version:
    Square with 13 mm grid
  • Height:
    15 mm
  • Pk:
    300

Specifications

About this item

Features 13 mm grid marked in both directions. Suitable for bacteriological typing, probe assays, colony counts, susceptibility testing or any research and bacterial culture work requiring a large surface area.