Radiation storage boxes, SP Bel-Art
Nome del fornitore: Bel-Art Products, a Part of SP
Certificati
Informazioni su questo articolo
Storage boxes, lead lined polyethylene, suitable for storing vials of isotopes, contaminated instruments and tools, radioactive samples, solutions, or waste.
- Safeguard personnel from exposure to radiation or lead
- Handle opening is 89×29 mm
It has 0,76 mm thick lead shield encased in 6,4 mm thick high-density polyethylene which helps contain radiation without lead exposure. Each box has a cover of the same lead/polyethylene construction with a 32 mm external overlap designed to prevent radiation escape.
Frequently Bought Together