Orders LinkContactUsLinkComponent
Combined pH electrode, InLab® Flow
Supplier:   Mettler - Toledo
Combined pH electrode, InLab® Flow
Supplier:  Mettler - Toledo
Certificates
Combined pH electrode, InLab® Flow
Combined pH electrode, InLab® Flow
3 Options Available Below
Product Details & Documents
A glass-body, combination pH electrode with SteadyForce™ and S7 screw head for inline measurements in flow-through cell 611.
  • SteadyForce™ reference system: Pressurised (3 bar) electrolyte ensures electrolyte flow even in viscous samples and guarantees highly reproducible results
  • Double ceramic junction: Ideal for general applications
  • Glass body is highly chemical resistant
 : Cable and flow-through cell need to be ordered separately.
Product Family Options
Product Information
  • Type
    pH
    Temp.
    Material
    Electrolyte
    Diaphr.
    Ø×L
    Connection
    For
    Availability
    Price
  • METN59902854
    Combined pH electrode, InLab® Flow
    Combined pH electrode, InLab® Flow
    U402-S7/120
    0 - 14
    0...+80 °C
    Glass
    DPA-gel
    2×ceramic
    12×120 mm
    S7
    Measurements in flow-through cell
    Log in for availability
    Log in for availability
    1 * 1 Un.
    1 * 1 Un.
  • 662-2092
    Combined pH electrode, InLab® Flow
    Combined pH electrode, InLab® Flow
    InLab® Flow
    1 - 11
    0...+80 °C
    Glass
    DPA-gel
    2×ceramic
    7×40 mm
    S7
    Measurements in flow-through cell
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 560.00 SFr.
    560.00 SFr.
    1 * 1 ST
       560.00 SFr.
This product has been blocked by your organization. Please contact your purchasing department for more information
Flow-through cell
Product Information
  • Type
    pH
    Temp.
    Material
    Electrolyte
    Diaphr.
    Ø×L
    Connection
    For
    Availability
    Price
  • METT59904354
    Flow-through cell 611, for InLab® Flow, thermostatable, measuring volume 0,5 ml
    Flow-through cell 611, for InLab® Flow, thermostatable, measuring volume 0,5 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
Accessories
Product Information
  • Catalog #
    Length
    Availability
    Price
  • InLab® cable S7-BNC
    InLab® cable S7-BNC
    661-0257
    1,2 m
    Log in for availability
    1 * 1 ST - SFr. 78.00
Product Information
  • Catalog #
    Length
    Availability
    Price
  • InLab® cable S7-4mm
    InLab® cable S7-4mm
    661-0264
    1,2 m
    Log in for availability
    1 * 1 ST - SFr. 86.00
Product Information
  • Catalog #
    Length
    Availability
    Price
  • InLab® cable S7-DIN
    InLab® cable S7-DIN
    661-0261
    1,2 m
    Log in for availability
    1 * 1 ST - SFr. 86.70
Product Information
  • Catalog #
    Length
    Availability
    Price
  • InLab® cable S7-2mm
    InLab® cable S7-2mm
    661-0265
    1,2 m
    Log in for availability
    1 * 1 ST - SFr. 86.00
Product Information
  • Catalog #
    Length
    Availability
    Price
  • InLab® cable S7-Lemo 00
    InLab® cable S7-Lemo 00
    661-0267
    1,2 m
    Log in for availability
    1 * 1 ST - SFr. 211.00
Product Information
  • Catalog #
    Length
    Availability
    Price
  • InLab® cable S7-RCA
    InLab® cable S7-RCA
    661-0266
    1,2 m
    Log in for availability
    1 * 1 ST - SFr. 84.00
Product Information
  • Catalog #
    Length
    Availability
    Price
  • InLab® cable S7-BNC IP67
    InLab® cable S7-BNC IP67
    661-0260
    1,2 m
    Log in for availability
    1 * 1 ST - SFr. 86.00