Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Nutrient medium, Biosart® 100

Nutrient medium, Biosart® 100
Supplier:  Sartorius
Certificates
Nutrient medium, Biosart® 100
Nutrient medium, Biosart® 100

13 Options Available Below

Product Details & Documents

These ready to use ampoules are pre-sterilised and contain a membrane filter and pad. They have been specially designed for performing microbiological analyses on pharmaceuticals, cosmetics, food and drink, water and other liquids. To be used with Biosart® 100 Monitors. When stored in appropriate conditions (4 °C), the medium has a shelf life of up to 12 months (except for Endo). The formula ensures optimum colony growth on membranes and complies with international standards, including International Pharmacopoeias, DIN and ISO standards, American Standards for Water and Foods, mineral water regulations and recommendations for the food and drink industry.

  • Pre-sterilised nutrient medium with quality certificate for each batch
  • Consistently high recovery rates
  • Ready for use and easy to handle, avoids use of glass ampoules
  • Long shelf life

 : Pre-sterilised Biosart® 100 nutrient medium in 2.5 ml plastic ampoules. Each pack contains 50 plastic ampoules and a batch quality certificate.

 

Product Family Options

Product Information

  • For
    Pk
    Availability
    Price
  • TSA (USP)
    Total colony count
    50 Ampoul
    Log in for availability
    Log in for availability
    1 * 50 Ampull
    1 * 50 Ampull - SFr. 163.00
    SFr. 163.00
    1 * 50 Ampull
       SFr. 163.00

  • m-Endo
    E. coli and coliforms
    50 Ampoul
    Log in for availability
    Log in for availability
    1 * 50 Ampull
    1 * 50 Ampull - SFr. 163.00
    SFr. 163.00
    1 * 50 Ampull
       SFr. 163.00

  • 516-7020
    Sabouraud (USP)
    Sabouraud (USP)
    Yeast and Moulds
    50 Ampoul
    Log in for availability
    Log in for availability
    1 * 50 Ampull
    1 * 50 Ampull - SFr. 163.00
    SFr. 163.00
    1 * 50 Ampull
       SFr. 163.00

  • SART16400-02-KF-K
    KF-Streptococcus agar
    KF-Streptococcus agar
    Faecal Streptococcus
    50 Ampoul
    Log in for availability
    Log in for availability
    1 * 50 Ampull
    1 * 50 Ampull - SFr. 135.00
    SFr. 135.00
    1 * 50 Ampull
       SFr. 135.00

  • R2A agar
    Total count
    50 Ampoul
    Log in for availability
    Log in for availability
    1 * 50 Ampull
    1 * 50 Ampull - SFr. 163.00
    SFr. 163.00
    1 * 50 Ampull
       SFr. 163.00

  • SART16400-02-CE-K
    Cetrimide agar
    Cetrimide agar
    Pseudomonas
    50 Ampoul
    Log in for availability
    Log in for availability
    1 * 50 Ampull
    1 * 50 Ampull - SFr. 163.00
    SFr. 163.00
    1 * 50 Ampull
       SFr. 163.00

  • SART16400-02-OS-K
    M Green yeast and mould
    M Green yeast and mould
    Yeast and Moulds
    50 Ampoul
    Log in for availability
    Log in for availability
    1 * 50 Ampull
    1 * 50 Ampull - SFr. 163.00
    SFr. 163.00
    1 * 50 Ampull
       SFr. 163.00

  • M-FC broth
    Faecal Coliforms
    50 Ampoul
    Log in for availability
    Log in for availability
    1 * 50 Ampull
    1 * 50 Ampull - SFr. 163.00
    SFr. 163.00
    1 * 50 Ampull
       SFr. 163.00

  • 710-0855
    Wallerstein nutrient
    Wallerstein nutrient
    Yeast, mould & bacteria
    50 Ampoul
    Log in for availability
    Log in for availability
    1 * 50 Ampull
    1 * 50 Ampull - SFr. 163.00
    SFr. 163.00
    1 * 50 Ampull
       SFr. 163.00

  • SART16400-02-MG-K
    M Green yeast and mould
    M Green yeast and mould
    Yeast and Moulds
    50 Ampoul
    Log in for availability
    Log in for availability
    1 * 50 Ampull
    1 * 50 Ampull - SFr. 163.00
    SFr. 163.00
    1 * 50 Ampull
       SFr. 163.00

  • TTC
    Total colony count
    50 Ampoul
    Log in for availability
    Log in for availability
    1 * 50 Ampull
    1 * 50 Ampull - SFr. 163.00
    SFr. 163.00
    1 * 50 Ampull
       SFr. 163.00

  • 710-0856
    Wallerstein differential
    Wallerstein differential
    Bacteria in fermentation processes
    50 Ampoul
    Log in for availability
    Log in for availability
    1 * 50 Ampull
    1 * 50 Ampull - SFr. 182.00
    SFr. 182.00
    1 * 50 Ampull
       SFr. 182.00

  • SART16400-02-GS-K
    M Green selective Biosart
    M Green selective Biosart
    Yeast and Moulds
    50 Ampoul
    Log in for availability
    Log in for availability
    1 * 50 Ampull
    1 * 50 Ampull - SFr. 166.00
    SFr. 166.00
    1 * 50 Ampull
       SFr. 166.00