Orders LinkContactUsLinkComponent
Maintenance sorbents
Supplier:   3M
Maintenance sorbents
Supplier:  3M
Certificates
Maintenance sorbents
Maintenance sorbents
3 Options Available Below
Product Details & Documents
Light grey when dry and darken when wet as an indication of saturation level.
  • Inert, synthetic fibres principally of PP and polyester
  • Multi-format combines four formats in one product and can be used as booms, pillows, sheets or rolls
  • Minibooms are designed for placing around the base of equipment to contain and control leaks and spills
  • Minipillows are high capacity for use at the source of a leak. They are tear resistant and can be left in place for long periods
Used for the control of accidental and routine leaks, drips and spills of oils, lubricants and other non aggressive liquids. They are perfect for cleaning up non aggressive fluids as well as being an effective maintenance management tool.
Product Family Options
Product Information
  • Absorbency
    Packed
    L×W
    Availability
    Price
  • 121-1001
    Multiformat
    Multiformat
    37 litres/kit
    Box (3) of 15,2 m roll
    15,2 m × 120 mm
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - SFr. 387.00
    SFr. 387.00
    1 * 3 ST
       SFr. 387.00
  • 121-0016
    Minibooms
    Minibooms
    45 litres/box
    Box of 12 minibooms
    1,2 m × Ø 76 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - SFr. 192.00
    SFr. 192.00
    1 * 12 ST
       SFr. 192.00
  • 129-0146
    Minipillows
    Minipillows
    32 litres/box
    Box of 16 minipillows
    380×180 mm
    Log in for availability
    Log in for availability
    1 * 16 ST
    1 * 16 ST - SFr. 136.00
    SFr. 136.00
    1 * 16 ST
       SFr. 136.00