Springen zu Kopfbereich
Springen zu Hauptinhalt
Springen zu Fußzeile
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Magnesia
Magnesia, rods , Ø×H 1,8×140 mm
Magnesia, rods , Ø×H 1,8×140 mm
Magnesia, rods , Ø×H 1,8×140 mm
Catalog # 459-2912
Supplier:
CERAM TEC
CAS Number:
Expand image
Magnesia, rods , Ø×H 1,8×140 mm
Catalog # 459-2912
Supplier:
CERAM TEC
CAS Number:
Specifications
Pk:
100
Seq:
0100
Specifications
About this item
Produktdetails
:
:
1309-48-4
Produktdetails