Boiling Stones
Supplier: POLLATH
Certificates
About this item
Grade A: For preparative work. Can be used in aqueous, neutral, basic media. Highly effective even in the case of multiple interruptions of the boiling process in vacuum distillation.
Grade B: For analytical work, with acidic, neutral, basic media. Jump after interruption again. Not suitable for vacuum distillation.
Frequently Bought Together