Sampling knives, sterile, Corning® Gosselin™
231-0058
: Corning
- Colour:White
- Description:Sampling tools, knives, sterile
- Total length:175 mm
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:RECs, ISO 14001 - Borre
- Pk:1.000
PS, white. Knife for taking samples, as well as a variety of other uses.
- For the removal of small, solid samples
- Irradiated, traceable by the printed expiry date on the bottom of the packaging
- Ideal for single use
: *The product(s) on this page are outside the scope of the EU Directive 2019/904