Round canisters
About this item
SCAT round canisters are ideal for collecting and disposing of smaller quantities of solvents due to their compact dimensions. Several SCAT round canisters can be easily stationed at the HPLC system and equipped with SCAT SafeyWasteCaps.
- Made of either robust HDPE or PP and therefore resistant to all solvents commonly used in the laboratory
- Available in the common threads S65, S70/71 and B83
- Filling capacities range from very compact 2 to 4 L, 10, 25, 50 and 60 L for the collection of voluminous solvents
- Many individual features such as carrying handles, handles or measuring scales, which ensure safe handling of the hazardous substances
SCAT round canisters are a perfect alternative or supplement to conventional SCAT canisters to provide physical separation of solvents and prevent mixing of hazardous liquids.
8 Options Available Below
Product Details & Documents
SCAT round canisters are ideal for collecting and disposing of smaller quantities of solvents due to their compact dimensions. Several SCAT round canisters can be easily stationed at the HPLC system and equipped with SCAT SafeyWasteCaps.
- Made of either robust HDPE or PP and therefore resistant to all solvents commonly used in the laboratory
- Available in the common threads S65, S70/71 and B83
- Filling capacities range from very compact 2 to 4 L, 10, 25, 50 and 60 L for the collection of voluminous solvents
- Many individual features such as carrying handles, handles or measuring scales, which ensure safe handling of the hazardous substances
SCAT round canisters are a perfect alternative or supplement to conventional SCAT canisters to provide physical separation of solvents and prevent mixing of hazardous liquids.
Product Family Options
Product Information
- CapacityColourThreadTypeW×D×HAvailabilityPrice
- SCTV107722Blue liter scaleBlue liter scale5 LNaturalS65with handle and
measuring scale167×167×330 mmLog in for availability1 * 1 STSFr. 142.00 - 590-0572Blue liter scaleBlue liter scale10 LNaturalS65With handle and
measuring scale205×205×430 mmLog in for availability1 * 1 STSFr. 174.00 - 590-0573with handlewith handle10 LNaturalB83With carrying handles, autoclavable250×250×390 mmLog in for availability1 * 1 STSFr. 200.00
- SCTV107720Blue liter scaleBlue liter scale25 LNaturalS65With measuring scale278×278×580 mmLog in for availability1 * 1 STSFr. 212.00
- 50 LNaturalB83With carrying handles, autoclavable380×380×680 mmLog in for availability1 * 1 STSFr. 438.00
- SCTV107721Blue liter scaleBlue liter scale60 LNaturalS65With measuring scale350×350×825 mmLog in for availability1 * 1 STSFr. 313.00



