WHEATON® Bottles, Wide Neck, with Screw Cap
Supplier: DWK Life Sciences
Certificates
About this item
Amber, USP Type III soda-lime glass.
- Easy removal of contents
- White polypropylene screw cap
- Hazardous communication label included with each case of bottles allows for content ID and safety notification
- Tamper-evident seal included with each case of bottles keeps bottle sealed which protecting contents from accidental misuse
Ideal for storage of light-sensitive samples or dry powders.