Sample spoons, Sterileware®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
PS, white. These economical spoons are ideal for retrieving small, specific volume samples of powders or liquids. Use a separate spoon for each material sampled to avoid cross contamination.
- Unique flat bottom shape allows them to be used as weighing boats for additional sampling accuracy
- Made from white, FDA grade high-impact PS for durability and stiffness when sampling packed powders
- Sterile spoons are individually sealed and sterilised in a polyethylene bag ready for one time use and disposal
- Lot stamped for accurate lot tracing and shipped with a Certificate of Processing for Sterilization
Frequently Bought Together