Organic reference standard, Sulfathiazole 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA17000200AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:72-14-0
- Description:Sulfathiazole 100 µg/ml in acetonitrile
- Formula:C₉H₉N₃O₂S₂
- Description:Sulfathiazole