Curcumin for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:458-37-7
:
Formula:C₂₁H₂₀O₆
MDL Number:MFCD00008365
:368.39 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:207-280-5
Specification Test Results
- Assay (HPLC, area%):≥ 75.0 % (a/a)
- Bisdemethoxycurcumin (HPLC; Area%):≤ 5.0 % (a/a)
- Demethoxycurcumin (HPLC; Area%):≤ 20.0 % (a/a)
- Identity (IR):passes test
Specification Test Results
Frequently Bought Together