Order Entry
Switzerland
ContactUsLinkComponent
J.T.Baker®, Boron Standard Solution, 10000 mg/L B in Water 2% Ammonium Hydroxide (from B(OH₃)) Standard for ICP
J.T.Baker®, Boron Standard Solution, 10000 mg/L B in Water 2% Ammonium Hydroxide (from B(OH₃)) Standard for ICP
Catalog # 5722.0100
Supplier:  Avantor
CAS Number:  
J.T.Baker®, Boron Standard Solution, 10000 mg/L B in Water 2% Ammonium Hydroxide (from B(OH₃)) Standard for ICP
Catalog # 5722.0100
Supplier:  Avantor
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    2% NH₄OH
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Boron
  • Symbol:
    B

Specifications

About this item

Certifications: Traceable to NIST