Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Melatonin

Melatonin

Glass bottle for solids, 250 mg
Catalog # ROTH6956.1
Marketsource
Supplier:   Roth Carl
Melatonin
Catalog # ROTH6956.1
Supplier:  Roth Carl
Marketsource
CAS Number:  73-31-4
Melatonin

Glass bottle for solids, 250 mg

CAS Number:  73-31-4
Price
SFr. 94.20
Log in for availability
1 * 250 mg
Product Details & Documents
Synonyms:  Melatonin, N-(2-(5-Methoxy-1H-indol-3-yl)ethyl)acetamide
  • CAS Number:73-31-4
  • Formula:C₁₃H₁₆N₂O₂
  • MDL Number:MFCD00005655
  • Molecular Weight:232.28 g/mol
  • EINECS:200-797-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.