Order Entry
Switzerland
ContactUsLinkComponent
Antimony standard solution, 1000 mg/L Sb in water standard for AAS
Antimony standard solution, 1000 mg/L Sb in water standard for AAS
Catalog # REGEAASBH
Supplier:  REAGECON
CAS Number:  
Antimony standard solution, 1000 mg/L Sb in water standard for AAS
Catalog # REGEAASBH
Supplier:  REAGECON
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Element:
    Antimony
  • Symbol:
    Sb

Specifications

About this item