Order Entry
Switzerland
ContactUsLinkComponent
Ethanol denatured absolute (untaxed) ≥99.8% (based on denaturant-free substance), Purum A15 T1 (denatured with 1% toluene)
Ethanol denatured absolute (untaxed) ≥99.8% (based on denaturant-free substance), Purum A15 T1 (denatured with 1% toluene)
Catalog # 02880-10L
CAS Number:  
Ethanol denatured absolute (untaxed) ≥99.8% (based on denaturant-free substance), Purum A15 T1 (denatured with 1% toluene)
Catalog # 02880-10L
CAS Number:  

Specifications

  • Pack type:
    Stainless steel can
  • Pk:
    10 L
  • Seq:
    5214

Specifications

Specification Test Results

  • Purity without addives GC):
    Min. 99.8 Gew.%
  • Refractive index (n 20/D):
    1.362 - 1.364
  • Non-volatile matter:
    Max. 0.001 %
  • Water (Karl Fischer):
    Max. 0.3 %
  • Toluene (GC):
    0.9 - 1.5 %
  • Autoignition Temperature:
    683 °F
  • Vapor Density:
    1.59 (vs air)
  • Explosion Limit:
    19 %, 60 °F
  • Refractive Index:
    n20/D 1.363

Specification Test Results

About this item

 :
 :46.07 g/mol
Boiling Pt:78 °C (1.013 hPa)
Melting Pt:–130 °C
Density:0.80 g/cm3
Flash Pt:9 °C
Storage Temperature:Ambient
EINECS:203-625-9
UN:1170
ADR:3,II
 :GHS07
 :GHS02
CAS Number:64-17-5
Formula:H₃CCH₂OH
MDL Number:MFCD00003568