Order Entry
Switzerland
ContactUsLinkComponent
Boron standard solution, 500 mg/L B in water (from B(OH₃))
Boron standard solution, 500 mg/L B in water (from B(OH₃))
Codice catalogo CMPLFC31.K5.W.L25
Nome del fornitore:  CUSTOM MADE CHEMICALS LAB
Numero CAS:  
Boron standard solution, 500 mg/L B in water (from B(OH₃))
Codice catalogo CMPLFC31.K5.W.L25
Nome del fornitore:  CUSTOM MADE CHEMICALS LAB
Numero CAS:  

Specifiche

  • Concentration:
    500 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    250 mL
  • Element:
    Boron
  • Symbol:
    B

Specifiche

Informazioni su questo articolo