SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Standards
Cationic Standards
Boron Standards
Boron standard solution, 400 mg/L B in water (from B(OH₃))
Boron standard solution, 400 mg/L B in water (from B(OH₃))
CMPLFC31.K4.W.L25
:
CUSTOM MADE CHEMICALS LAB
:
Boron standard solution, 400 mg/L B in water (from B(OH₃))
CMPLFC31.K4.W.L25
:
CUSTOM MADE CHEMICALS LAB
:
Concentration:
400 mg/L
Matrix:
H₂O
Pack type:
Plastic bottle
Pk:
250 mL
Element:
Boron
Symbol:
B