Order Entry
Switzerland
ContactUsLinkComponent
di-Lithium oxalate for synthesis, Sigma-Aldrich®
Irritant
di-Lithium oxalate for synthesis, Sigma-Aldrich®
Catalog # 8.22085.0100
Supplier:  Merck
CAS Number:  
Irritant
di-Lithium oxalate for synthesis, Sigma-Aldrich®
Catalog # 8.22085.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Solubility:
    66 g/l (20 °C)
  • Molar Mass:
    101.9 g/mol

Specification Test Results

About this item

 :
 :101.9 g/mol
ADR:6.1,III
 :M-Clarity™ quality level = MQ200
EINECS:209-054-1
UN:3288
 :GHS07
Storage Temperature:Ambient
CAS Number:553-91-3
 :30903-87-8
Formula:C₂Li₂O₄
MDL Number:MFCD00040596