Pyridoxal hydrochloride ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:65-22-5
:
Formula:C₈H₁₀ClNO₃
MDL Number:MFCD00012809
:203.62 g/mol
Merck Index:14,7978
Melting Pt:172…174 °C (dec.)
EINECS:200-602-5
Related Information
Frequently Bought Together