Pyridoxal hydrochloride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:65-22-5
:
Formula:C₈H₁₀ClNO₃
MDL Number:MFCD00012809
:203.62 g/mol
Melting Pt:171…175 °C (dec.)
Storage Temperature:Refrigerator
EINECS:200-602-5
Specification Test Results
- Appearance:White to yellow to light beige Powder
- Infrared spectrum:Conforms
- Melting point:171 to 175 °C (Dec)
- Titration with HClO4:≥98.5 %
Specification Test Results
Frequently Bought Together