Order Entry
Switzerland
ContactUsLinkComponent
Acetic acid ≥99.0%, TraceSELECT™ for trace analysis, Fluka™
Acetic acid ≥99.0%, TraceSELECT™ for trace analysis, Fluka™
Certificates
undefined
Acetic acid ≥99.0%, TraceSELECT™ for trace analysis, Fluka™

Specification Test Results

  • assay (HPLC):
    Min. 99 %
  • measurement uncertainty:
    effective %
  • identity (NMR):
    complying
  • melting range:
    62 - 78 °C
  • Barium (Ba):
    Max. 0.5 ppb
  • Beryllium (Be):
    Max. 0.5 ppb
  • loss on drying (105°C, 5h):
    15 - 30 %
  • Calcium (Ca):
    Max. 20 ppb
  • Cadmium (Cd):
    Max. 0.5 ppb
  • Cerium (Ce):
    Max. 0.5 ppb
  • Cobalt (Co):
    Max. 0.5 ppb
  • Chromium (Cr):
    Max. 1 ppb
  • Cesium (Cs):
    Max. 0.5 ppb
  • Copper (Cu):
    Max. 2 ppb
  • Iron (Fe):
    Max. 10 ppb
  • Gallium (Ga):
    Max. 0.5 ppb
  • Germanium (Ge):
    Max. 0.5 ppb
  • Mercury (Hg):
    Max. 1 ppb
  • Indium (In):
    Max. 0.5 ppb
  • Potassium (K):
    Max. 10 ppb
  • Lithium (Li):
    Max. 0.5 ppb
  • Magnesium (Mg):
    Max. 5 ppb
  • Manganese (Mn):
    Max. 0.5 ppb
  • Molybdenum (Mo):
    Max. 0.5 ppb
  • Sodium (Na):
    Max. 20 ppb
  • Nickel (Ni):
    Max. 1 ppb
  • Lead (Pb):
    Max. 1 ppb
  • Platinum (Pt):
    Max. 0.5 ppb
  • Rubidium (Rb):
    Max. 0.5 ppb
  • Rhodium (Rh):
    Max. 0.5 ppb
  • Ruthenium (Ru):
    Max. 0.5 ppb
  • Antimony (Sb):
    Max. 0.5 ppb
  • Selenium (Se):
    Max. 1 ppb
  • Tin (Sn):
    Max. 0.5 ppb
  • Strontium (Sr):
    Max. 0.5 ppb
  • Titanium (Ti):
    Max. 1 ppb
  • Thallium (Tl):
    Max. 0.5 ppb
  • Uran (U):
    Max. 0.5 ppb
  • Vanadium (V):
    Max. 1 ppb
  • Zinc (Zn):
    Max. 2 ppb
  • Zirconium (Zr):
    Max. 0.5 ppb
  • Refractive Index:
    n20/D 1.371 (lit.)
  • Explosion Limit:
    16 %, 92 °F

Specification Test Results

Related Information

Frequently Bought Together