Order Entry
Switzerland
ContactUsLinkComponent
 
Orthophosphoric acid 85% ≥85%, CMOS for microelectronic, J.T.Baker®
  0264-05
 :  Avantor
 :  
undefined
Orthophosphoric acid 85% ≥85%, CMOS for microelectronic, J.T.Baker®
  0264-05
 :  Avantor
 :  

 

  • Pk:
    14 lb
  • Seq:
    7104
  • Environmentally Preferable:

 

 

  • Assay (H₃PO₄) (by acidimetry) :
    85.0 - 87.0 %
  • Color (APHA) :
    ≤ 10 5
  • Specific Gravity at 60°/60°F :
    1.691 - 1.710
  • Reducing Substances:
    Passes Test
  • Volatile Acids (µeq/g) :
    ≤ 0.16
  • Chloride (Cl) :
    ≤ 1 ppm
  • Nitrate (NO₃):
    ≤ 2 ppm
  • Sulfate (SO₄) :
    ≤ 12 ppm
  • Trace Impurities - Aluminum (Al) :
    ≤ 0.500 ppm
  • Trace Impurities - Antimony (Sb) :
    ≤ 10.000 ppm
  • Trace Impurities - Arsenic (As) :
    ≤ 0.050 ppm
  • Trace Impurities - Calcium (Ca) :
    ≤ 1.500 ppm
  • Trace Impurities - Chromium (Cr) :
    ≤ 0.200 ppm
  • Trace Impurities - Cobalt (Co) :
    ≤ 0.050 ppm
  • Trace Impurities - Copper (Cu) :
    ≤ 0.050 ppm
  • Trace Impurities - Gold (Au) :
    ≤ 0.300 ppm
  • Trace Impurities - ACS - Heavy Metals (as Pb) :
    ≤ 5 ppm
  • Trace Impurities - Iron (Fe) :
    ≤ 2.000 ppm
  • Trace Impurities - Lead (Pb) :
    ≤ 0.300 ppm
  • Trace Impurities - Lithium (Li) :
    ≤ 0.100 ppm
  • Trace Impurities - Magnesium (Mg) :
    ≤ 0.20 ppm
  • Trace Impurities - Manganese (Mn) :
    ≤ 0.100 ppm
  • Trace Impurities - Nickel (Ni) :
    ≤ 0.200 ppm
  • Trace Impurities - Potassium (K) :
    ≤ 1.500 ppm
  • Trace Impurities - Sodium (Na) :
    ≤ 2.500 ppm
  • Trace Impurities - Strontium (Sr) :
    ≤ 0.100 ppm
  • Trace Impurities - Titanium (Ti) :
    ≤ 0.300 ppm
  • Trace Impurities - Zinc (Zn) :
    ≤ 2.000 ppm
  • Particle Count - 1.0 µm and greater :
    ≤ 10 par/ml
  • Storage Conditions:
    Store at temperatures above 22°C (72°F)

 

 

 : Phosphoric acid
 :
 :98 g/mol
 :28 °C
 :1.71…1.87 g/cm³ (25 °C)
 :Ambient
 :231-633-2
 :1805
 :8,III
 :GHS05
 :7664-38-2
 :H₃PO₄
 :MFCD00011340