Order Entry
Switzerland
ContactUsLinkComponent
Boron standard solution, 1,000 mg/L B in water standard for ICP
Boron standard solution, 1,000 mg/L B in water standard for ICP
Catalog # REGEPB2A7
Supplier:  REAGECON
CAS Number:  
Boron standard solution, 1,000 mg/L B in water standard for ICP
Catalog # REGEPB2A7
Supplier:  REAGECON
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Boron
  • Symbol:
    B

Specifications

About this item