Boron standard solution, 1000 mg/L B in water (from B(OH₃)), standard for AAS, Specpure®
Molecular Formula: H₃BO₃ in H₂O
Physical Form: Liquid
MDL No.: MFCD00134034
: Traceable to NIST.
- MDL No.:MFCD00134034
Frequently Bought Together
Molecular Formula: H₃BO₃ in H₂O
Physical Form: Liquid
MDL No.: MFCD00134034