Order Entry
Switzerland
ContactUsLinkComponent
 
J.T.Baker®, Beryllium Standard Solution, 10000 mg/L Be in dil. Nitric Acid (from BeO.(Be(OAc)₂)₃) Standard for ICP
  5720.0100
 :  Avantor
 :  
J.T.Baker®, Beryllium Standard Solution, 10000 mg/L Be in dil. Nitric Acid (from BeO.(Be(OAc)₂)₃) Standard for ICP
  5720.0100
 :  Avantor
 :  

 

  • Concentration:
    10000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Beryllium
  • Symbol:
    Be

 

 

 : Traceable to NIST.