

Hydrochloric acid 36.5% - 38.0% NF, FCC, ACS, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–30 °C
Density:1.18 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:8,II
:Management of Change (MOC) category = R
:36.46 g/mol
EINECS:231-595-7
UN:1789
:GHS05
:GHS07
Specification Test Results
- Bulk Food Chemical:36.5 - 38.0 %
- Bulk Pharmaceutical Chemical:Passes Test
- NF - Assay (HCl)(NF):36.5 - 38.0 %
- NF - Bromide or Iodide:Passes Test
- NF - Free Bromine or Chlorine:Passes Test
- NF - Identification:Passes Test
- NF - Residue on Ignition:≤ 0.008 %
- NF - Sulfate (SO₄):Passes Test
- NF - Sulfite (SO₃):Passes Test
- FCC - Assay (HCl)(FCC):36.5 - 38.0 %
- FCC - Color (APHA):Passes Test
- FCC - Baume Degrees:22.7 - 23.4
- FCC - Identification:Passes Test
- FCC - Iron (Fe) (mg/kg):≤ 5
- FCC - Lead (Pb) (mg/kg):≤ 1
- FCC - Mercury (Hg)(mg/kg):≤ 0.10
- FCC - Nonvolatile Residue:≤ 0.5 %
- Total Organic Compounds - Non-Fluorine Containing (mg/kg):≤ 5
- Total Organic Compounds - Benzene (mg/kg):≤ 0.05
- Total Organic Compounds - Fluorinated Organic Compounds(mg/kg):≤ 25
- Total Organic Compounds - Oxidizing Substances:≤ 0.003 %
- Reducing Substances:≤ 0.007 %
- Specific Gravity:1.1856 - 1.1924
- Sulfate (SO₄):≤ 0.5 %
- ACS - Assay (HCl):36.5 - 38.0 %
- ACS - Ammonium (NH₄):≤ 3 ppm
- ACS - Arsenic (As):≤ 0.010 ppm
- ACS - Bromide (Br):≤ 0.005 %
- ACS - Color (APHA):≤ 10
- ACS - Extractable Organic Substances:≤ 5 ppm
- ACS - Free Chlorine (as Cl):≤ 1 ppm
- ACS - Heavy Metals (as Pb):≤ 1 ppm
- Trace Impurities - ACS - Iron (Fe):≤ 0.200 ppm
- ACS - Residue after Ignition:≤ 5 ppm
- ACS - Sulfate (SO₄):≤ 1 ppm
- ACS - Sulfite (SO₃):≤ 1 ppm
- Appearance:Passes Test
Specification Test Results
Related Information
Certificates
Search by Lot Numbersds_MACR2062
SDS
spec_MACR2062-04 spec_MACR2062-46
Product specification
Frequently Bought Together