ContactUsLinkComponent
VWR®, Slide Box
VWR®, Slide Box
Catalog # 631-9113
Supplier:  VWR Collection
undefined
VWR®, Slide Box
Catalog # 631-9113
Supplier:  VWR Collection
Specifications
  • No. of slides:
    100
  • Colour:
    Grey
  • Description:
    Slide box
  • W×D×H:
    208×175×34  mm
  • Pk:
    1
Specifications
About this item
This heavy-duty two-piece microscope slide box stores up to 100 slides.
  • Each slot is numbered to correspond to the slide inventory sheet on the inside cover of the box
  • Moulded, rigid box: Securely holds slides in grooved slots
  • Stackable for long-term storage
The box is made of durable ABS and secured with a rust resistant, nickel plated clasp and hinge pin for storage or transport.