Orders LinkContactUsLinkComponent
Cryoboxes
 :   RATIOLAB
Cryoboxes
 :  RATIOLAB
Cryoboxes
Cryoboxes
Cryoboxes, cardboard, for storage of cryovials at low temperatures.
  • Waterproof coating
  • Dividers must to be ordered separately
  • Available in different heights and five colours
Product Family Options
Cryoboxes
  • Compartments
    For
    Colour
    W×D×H ext.
  • -
    -
    White
    75×75×50 mm
    1 * 10 ST
    1 * 10 ST - SFr. 53.10
    SFr. 53.10
    1 * 10 ST
       SFr. 53.10
Dividers
  • Compartments
    For
    Colour
    W×D×H ext.
  • 5×5
    Tubes 11,0 mm Ø
    White
    68×68×30 mm
    1 * 10 ST
    1 * 10 ST - SFr. 24.70
    SFr. 24.70
    1 * 10 ST
       SFr. 24.70