ContactUsLinkComponent
VWR®, Cleanroom Disposable Goggles
VWR®, Cleanroom Disposable Goggles
Catalog # 111-0615
Supplier:  VWR Collection
undefined
VWR®, Cleanroom Disposable Goggles
Catalog # 111-0615
Supplier:  VWR Collection
Specifications
  • Type:
    Cleanroom, sterile, anti-fog, anti-scratch
  • Colour:
    Transparent
  • Lenses:
    Clear PC
  • Pk:
    10
Specifications
About this item
Sterile cleanroom goggles with a lightweight frame, indirect ventilation, and a non linting, latex-free headband.
  • Toughened PC lens with anti-fog and anti-scratch coating
  • Lightweight PVC frame with silicone rubber strap provides comfort
  • Ethylene oxide (EtO) sterilised and fully validated to ISO 11135-1-2007
Recommended for ISO Class 4 and EU GMP Grade A/B critical environments.
Certifications: EN 166:2001, Optical Class 1.
Packaging: In a Nextech certified ISO Class 4 for cleanroom. One piece per Tyvek™ PE bag.