Orders LinkContactUsLinkComponent
Stopcocks, miniature
Stopcocks, miniature
Supplier:   Bohlender
Stopcocks, miniature
Supplier:  Bohlender
Certificates
Stopcocks, miniature
Stopcocks, miniature
12 Options Available Below
Product Details & Documents
Miniature stopcocks of PTFE with male or female connection threads and stopcock plug of FEP.
  • Temperature resistance from -200 to 205°C
  • Universally chemical resistant
  • Pressure upto 8 bar
Product Family Options
2-way stopcocks, straight
Product Information
  • For tube I-Ø
    For tube O-Ø
    Thread
    Availability
    Price
  • BOHLF730-04
    Miniature stopcocks with two female thread connection
    Miniature stopcocks with two female thread connection
    1,6 mm
    3,2 mm
    UNF ¼" 28 G (male or female thread)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 80.80
    SFr. 80.80
    1 * 1 ST
       SFr. 80.80
  • BOHLF730-06
    Miniature stopcocks with a female thread and a male thread connection
    Miniature stopcocks with a female thread and a male thread connection
    0,8 mm
    1,6 mm
    UNF ¼" 28 G (male or female thread)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 217.00
    SFr. 217.00
    1 * 1 ST
       SFr. 217.00
  • BOHLF730-08
    Miniature stopcocks with a female thread and a male thread connection
    Miniature stopcocks with a female thread and a male thread connection
    1,6 mm
    3,2 mm
    UNF ¼" 28 G (male or female thread)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 130.00
    SFr. 130.00
    1 * 1 ST
       SFr. 130.00
  • BOHLF730-02
    Miniature stopcocks with two female thread connection
    Miniature stopcocks with two female thread connection
    0,8 mm
    1,6 mm
    UNF ¼" 28 G (male or female thread)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 85.30
    SFr. 85.30
    1 * 1 ST
       SFr. 85.30
3-way stopcocks, L-shaped bore
Product Information
  • For tube I-Ø
    For tube O-Ø
    Thread
    Availability
    Price
  • BOHLF731-02
    Miniature stopcocks with three female thread connection
    Miniature stopcocks with three female thread connection
    0,8 mm
    1,6 mm
    UNF ¼" 28 G (male or female thread)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 141.00
    SFr. 141.00
    1 * 1 ST
       SFr. 141.00
  • BOHLF731-04
    Miniature stopcocks with three female thread connection
    Miniature stopcocks with three female thread connection
    1,6 mm
    3,2 mm
    UNF ¼" 28 G (male or female thread)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 99.80
    SFr. 99.80
    1 * 1 ST
       SFr. 99.80
  • BOHLF731-12
    Miniature stopcocks with two female thread and a male thread connection
    Miniature stopcocks with two female thread and a male thread connection
    1,6 mm
    3,2 mm
    UNF ¼" 28 G (male or female thread)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 192.00
    SFr. 192.00
    1 * 1 ST
       SFr. 192.00
  • BOHLF731-10
    Miniature stopcocks with two female thread and a male thread connection
    Miniature stopcocks with two female thread and a male thread connection
    0,8 mm
    1,6 mm
    UNF ¼" 28 G (male or female thread)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 238.00
    SFr. 238.00
    1 * 1 ST
       SFr. 238.00
3-way stopcocks, T-shaped bore
Product Information
  • For tube I-Ø
    For tube O-Ø
    Thread
    Availability
    Price
  • BOHLF731-06
    Miniature stopcocks with three female thread connection
    Miniature stopcocks with three female thread connection
    0,8 mm
    1,6 mm
    UNF ¼" 28 G (male or female thread)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 111.00
    SFr. 111.00
    1 * 1 ST
       SFr. 111.00
  • BOHLF731-08
    Miniature stopcocks with three female thread connection
    Miniature stopcocks with three female thread connection
    1,6 mm
    3,2 mm
    UNF ¼" 28 G (male or female thread)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 97.30
    SFr. 97.30
    1 * 1 ST
       SFr. 97.30
  • BOHLF731-16
    Miniature stopcocks with two female thread and a male thread connection
    Miniature stopcocks with two female thread and a male thread connection
    1,6 mm
    3,2 mm
    UNF ¼" 28 G (male or female thread)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 256.00
    SFr. 256.00
    1 * 1 ST
       SFr. 256.00
  • BOHLF731-14
    Miniature stopcocks with two female thread and a male thread connection
    Miniature stopcocks with two female thread and a male thread connection
    0,8 mm
    1,6 mm
    UNF ¼" 28 G (male or female thread)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 333.00
    SFr. 333.00
    1 * 1 ST
       SFr. 333.00