ContactUsLinkComponent
Cleanroom gloves, nitrile, sterile, BioClean Nitramax™
Cleanroom gloves, nitrile, sterile, BioClean Nitramax™
Supplier:  Nitritex
Certificates
undefined
Cleanroom gloves, nitrile, sterile, BioClean Nitramax™
Supplier:  Nitritex
Certificates
Specifications
  • Catalog No:
  • 112-3859
  • 112-3860
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Cleanroom class:
  • 10/ISO 4; EU GMP (A)
  • 10/ISO 4; EU GMP (A)
  • Cuff style:
  • Beaded
  • Beaded
  • Cuff thickness:
  • 0,09 mm
  • 0,09 mm
  • Finger thickness:
  • 0,18 mm
  • 0,18 mm
  • Palm thickness:
  • 0,15 mm
  • 0,15 mm
  • EN norm:
  • EN 455, EN 374
  • EN 455, EN 374