ContactUsLinkComponent
BOD bottles, with robotic stopper, WHEATON®
BOD bottles, with robotic stopper, WHEATON®
Catalog # WHEA227667
Supplier:  DWK Life Sciences
undefined
BOD bottles, with robotic stopper, WHEATON®
Catalog # WHEA227667
Supplier:  DWK Life Sciences
Specifications
  • Capacity:
    300 ml
  • Description:
    BOD bottle, black, with PVC plastic coating, with glass robotic stopper, 73×167 mm (Ø×H)
  • Pk:
    20
Specifications
About this item
Made of Type I borosilicate glass, black.
  • For light sensitive samples
  • With glass Robotic stopper
  • Additional opaque black cap
This is a standard 300 ml BOD bottle with black PVC plastic coating that blocks all visible light up to 800 nm. This inhibits the production of oxygen by algae. We recommend this bottle for use in marine photosynthesis projects in which you need to compare oxygen in a light and a dark bottle.