dNTP sets, illustra™
Supplier: Cytiva
Jump to:
About this item
High purity dNTPs for amplification, dideoxy sequencing, labelling, mutagenesis, cDNA synthesis, and expression profiling.
- Free from DNase, RNase, and nicking enzyme activity
- Greater than 99% triphosphate purity (by HPLC) for consistent, high performance
- Buffer-free, ready to use solutions at a variety of concentrations
- Functionally tested to produce a 20,7 kb PCR amplification product from lambda DNA
5 Options Available Below
Product Details & Documents
High purity dNTPs for amplification, dideoxy sequencing, labelling, mutagenesis, cDNA synthesis, and expression profiling.
- Free from DNase, RNase, and nicking enzyme activity
- Greater than 99% triphosphate purity (by HPLC) for consistent, high performance
- Buffer-free, ready to use solutions at a variety of concentrations
- Functionally tested to produce a 20,7 kb PCR amplification product from lambda DNA
Documents
Certificates
Search by Lot Number28406552_HAND BOOK 28406552_INSTRUCTIONS 28406553_HAND BOOK 28406558_HAND BOOK 28406558_INSTRUCTIONS
Manual
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Product Family Options
Product Information
- PkAvailabilityPrice
- 28-4065-51dNTP set (100 mM each A,C,G,T), 4×25 µmol
- 28-4065-57dNTP mix set (20 mM A,C,G,T), 10 µmol
- 28-4065-52dNTP set (100 mM each A,C,G,T), 4×100 µmol
- 28-4065-53illustra™ dNTP set, 4×500 µmol (100 mM each A,C,G,T)illustra™ dNTP set, 4×500 µmol (100 mM each A,C,G,T)1 SETLog in for availability1 * 1 SET3 270.00 SFr.
- 28-4065-58dNTP mix set (20 mM A,C,G,T), 4×10 µmol
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



