ContactUsLinkComponent
Capillary cutter, CleanCut
Capillary cutter, CleanCut
Catalog # 590-0850
Supplier:  S.C.A.T.
undefined
Capillary cutter, CleanCut
Catalog # 590-0850
Supplier:  S.C.A.T.
Specifications
  • Description:
    CleanCut capillary tubing cutter, with replacement blade
  • Pk:
    1
Specifications
About this item
This cutter is made of PP, blue, with stainless steel blade.
  • Separates capillary tubes safely and reliably
  • With useful openings for measuring the capillary diameter
  • Prevents injuries and ensures a clean cut
The universal tube cutter for HPLC capillaries - the tube guide enables a comfortable, straight cut. With spare blade for exchange.