Snap caps for sample bottles, WHEATON®
Anbieter: DWK Life Sciences
Zertifikate
Über diesen Artikel
Natural low density polyethylene.
- All sizes have tabs for easy removal
- Good chemical resistance
- Good temperature resistance
For WHEATON snap cap sample bottles.
Frequently Bought Together