Reagent bottles, with screw cap, WHEATON®
Nome del fornitore: DWK Life Sciences
Certificati
Informazioni su questo articolo
Manufactured from USP Type I borosilicate glass, with large writing patch.
- Features no-drip pour lip
- Pre-attached polypropylene screw cap with PTFE liner
- Autoclavable
Frequently Bought Together